Skip to Content
Merck
All Photos(1)

Key Documents

58158AST

Supelco

CHIRALPAK® AGP (5 μm) HPLC Columns

L × I.D. 1 cm × 3 mm, HPLC Guard Cartridge, pk of 2 ea

Sign Into View Organizational & Contract Pricing

Select a Size

2 EA
$1,420.00

$1,420.00


Check Cart for Availability


Select a Size

Change View
2 EA
$1,420.00

About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

$1,420.00


Check Cart for Availability

Product Name

CHIRALPAK® AGP HPLC Guard Column, 5 μm particle size, L × I.D. 1 cm × 3 mm, pk of 2 ea

material

stainless steel column

Quality Level

Agency

suitable for USP L41

product line

CHIRALPAK®

packaging

pk of 2 ea

manufacturer/tradename

CHIRALPAK®

parameter

137 bar pressure (2000 psi)
20-30 °C temperature
40 °C max. temp.

technique(s)

HPLC: suitable

L × I.D.

1 cm × 3 mm

matrix

fully porous particle

matrix active group

glycoprotein, alpha1- phase

particle size

5 μm

operating pH

2-8

separation technique

chiral

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

CHIRALPAK is a registered trademark of Daicel Corp.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service