Skip to Content
Merck
All Photos(3)

Key Documents

57908-U

Supelco

Nitinol-core (NIT) SPME Fiber Assembly

greener alternative

CAR/PDMS, for use with autosampler or manual holder, Nitinol-core (NIT), 24 ga, 75 μm

Synonym(s):

SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS)

Sign Into View Organizational & Contract Pricing

Select a Size

3 EA
$1,130.00

$1,130.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
3 EA
$1,130.00

About This Item

UNSPSC Code:
41104008
NACRES:
NB.21

$1,130.00


Check Cart for Availability

Request a Bulk Order

Product Name

Nitinol-core (NIT) SPME Fiber Assembly Carboxen/Polydimethylsiloxane (Carboxen/PDMS), df 75 μm(CAR/PDMS, for use with autosampler or manual holder, needle size 24 ga

material

black hub (metallic)
metal alloy fiber

needle size

24 ga

packaging

pkg of 3 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

75 μm (CAR/PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

CAR/PDMS coating

compatibility

(For gasses and low molecular weight compounds (MW 30 - 225).)
for use with autosampler or manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

For Gases and Low Molecular Weight Compounds (MW 30-225)
Nitinol-core (NIT) SPME fibers were specifically engineered to be more reproducible and more robust than traditional SPME fibers that contain a fused silica (FS) core. The NIT fibers offer > 30% improvement in lot to lot and fiber to fiber reproducibility when compared to the FS versions, but offer equivalent selectivity for easy transfer of methods from FS to the more robust, reproducible NIT version.
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

SPME fibers with CAR/PDMS and PDMS/DVB coatings on nitinol core assembly enhance fiber performance for solid phase microextraction.

A comparison between Carboxen®/PDMS coated nitinol core and fused silica core adsorbent SPME fibers for factors, such as coating process, testing process, inertness, raw materials, and durability.

Protocols

High performance, reliability, and reproducibility of HS-SPME in combination with GC/MS for the determination of VOCs in water was proven in an interlaboratory trial. The new ISO 17943 using HS-SPME is an improvement on existing official methods for this determination in terms of sensitivity and selectivity.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service