Skip to Content
Merck
All Photos(1)

Key Documents

577550-U

Supelco

BIOshell A160 Peptide Phenyl-Hexyl (2.7 μm) HPLC Columns

L × I.D. 5 cm × 1 mm, HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$1,360.00

$1,360.00


Check Cart for Availability


Select a Size

Change View
50 MG
$1,360.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

$1,360.00


Check Cart for Availability

Product Name

BIOshell A160 Peptide Phenyl-Hexyl, 2.7 μm HPLC Column, column L × I.D. 5 cm × 1 mm

material

stainless steel hardware

Quality Level

Agency

suitable for USP L11

product line

BIOshell

feature

endcapped

packaging

pkg of 1 unit

extent of labeling

4.7% Carbon loading

technique(s)

HPLC: suitable

column L × I.D.

5 cm × 1 mm

surface area

90 m2/g

matrix

Fused-Core® particle platform

matrix active group

Phenyl-Hexyl (dimethylphenyl-hexylsilane) bonding phase

particle size

2.7 μm

pore size

160 Å pore size

operating pH range

2-9

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell A160 Peptide Phenyl-Hexyl is a high-speed, high-performance liquid chromatography column based on a 160 Å Fused-Core® particle design. The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency for high molecular weight solutes (up to 20 kDa) due to the shallow diffusion paths in the 0.5 μm thick porous shell and the small overall particle size of 2.7 μm. The end-capped, phenyl-hexyl bonded phase of the Peptide Phenyl-Hexyl provides a stable, reversed phase packing with a pore structure and pore size that is optimized for reversed-phase HPLC separations of peptides and polypeptides, using typical acidic mobile phases favored for proteomic applications.

Analysis Note

The target analytes include peptides and polypeptides < 20 kDa.

Legal Information

Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service