Skip to Content
Merck
All Photos(1)

Key Documents

577430-U

Supelco

BIOshell IgG 1000 Å Diphenyl (2.7 μm) HPLC Columns

L × I.D. 15 cm × 4.6 mm, HPLC Column

Synonym(s):

BIOshell IgG 1000 Å Diphenyl, 2.7 μm HPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,680.00

$1,680.00


Check Cart for Availability


Select a Size

Change View
1 EA
$1,680.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,680.00


Check Cart for Availability

Product Name

BIOshell IgG 1000 Å Diphenyl, 2.7 μm HPLC Column, column L × I.D. 15 cm × 4.6 mm

material

stainless steel column

Agency

suitable for USP L11

product line

BIOshell

feature

endcapped

packaging

1 ea of

extent of labeling

1.0% loading

technique(s)

HPLC: suitable
LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

column L × I.D.

15 cm × 4.6 mm

surface area

22 m2/g

matrix

Fused-Core® particle platform
superficially porous particle

matrix active group

bonding phase, Diphenyl (diphenylmethyl)

particle size

2.7 μm

pore size

1000 Å pore size

operating pH range

2-9

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

BIOshell IgG 1000 Å Diphenyl is a high-speed, high-performance liquid chromatography column based on a new wide-pore (1000 Å) Fused-Core® particle design. The particle provides a thin porous shell of high-purity silica surrounding a solid silica core. This particle design exhibits very high column efficiency due to the shallow diffusion paths in the 0.5 μm thin porous shell and the small overall particle size of 2.7 μm. The densely bonded, extensively endcapped diphenylmethyl bonded phase of BIOshell IgG 1000 Å Diphenyl provides a stable, reversed-phase packing that can be used for separating high molecular weight compounds, such as proteins.

Analysis Note

Target analytes include: monoclonal antibodies, antibody-drug conjugates, antibody fragments and large proteins with molecular weights up to 500 kDa.

Legal Information

Fused-Core is a registered trademark of Advanced Materials Technology, Inc.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service