Key Documents
57645
Sample Loop for 9010, 9125, and 9725 Injectors
volume 200 μL, PEEK
Select a Size
1 EA
$137.00
$137.00
Estimated to ship on29 May 2025
Request a Bulk Order
Select a Size
Change View
1 EA
$137.00
About This Item
UNSPSC Code:
41115854
$137.00
Estimated to ship on29 May 2025
Request a Bulk Order
material
PEEK
O.D.
1.6 mm
1/16 in.
volume
200 μL
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)