Skip to Content
Merck
All Photos(1)

Key Documents

575625-U

Supelco

Discovery® DSC-CN SPE 96-well Plate

bed wt. 50 mg/well, pk of 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104018

composition

bed wt., 50 mg/well

packaging

pk of 1

technique(s)

solid phase extraction (SPE): suitable

matrix active group

cyano bonding

application(s)

food and beverages

separation technique

normal phase
reversed phase

Looking for similar products? Visit Product Comparison Guide

General description

Retention Mechanism: Reversed-phase or Normal phase
Sample Matrix Compatibility: Aqueous solutions (biological fluids, water) when used in reversed-phase; or organic solvents, oils, and lipids when used in normal phase

  • Monomerically bonded, cyanopropyl (7 %C), endcapped
  • Can be used in either reversed-phase or normal phase
  • Ideal for very hydrophobic analytes that may be irreversibly retained on more hydrophobic sorbents such as DSC-18
  • Less retentive than DSC-Si or DSC-Diol when used in normal phase (organic matrices such as hexane or oils)
  • Allows for the rapid release of very polar molecules irreversibly retained on very polar sorbent

Legal Information

Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service