Key Documents
57463
Rheodyne® Model 3725i Preparative Sample Injector
stainless steel
Select a Size
1 EA
$3,240.00
$3,240.00
Check Cart for Availability
Request a Bulk Order
Select a Size
Change View
1 EA
$3,240.00
About This Item
UNSPSC Code:
41115854
$3,240.00
Check Cart for Availability
Request a Bulk Order
material
stainless steel
feature
manual
number of ports 6
positions 2
packaging
pkg of 1 ea
General description
Model 3725i sample injectors for 1-10 cm I.D. HPLC columns combine the ease of use and versatility of Rheodyne′s analytical-scale injectors with the ability to handle large samples and high flow rates. Rugged and easily maintained, in a choice of stainless steel or PEEK flowpaths. Includes 10 mL sample loop, 1/8 in. fittings for all ports, 5cm large-bore syringe needle (16 gauge/0.65 in. OD) for rapid injection of large samples, needle port cleaner, two vent tubes, wrenches, mounting screws, and instructions.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Legal Information
Rheodyne is a registered trademark of IDEX Health & Science, LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)