Skip to Content
Merck
All Photos(1)

Documents

57332

Supelco

Thermometer

L 5 in., temperature -10-110 °C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

material

glass fiber

parameter

-10-110 °C temperature

technique(s)

solid phase microextraction (SPME): suitable

L

5 in.

application(s)

food and beverages

compatibility

for use with SPME sampling stand

General description

For monitoring sample temperature when using the SPME sampling stand and a heat/stir plate.

Application

Thermometer was used in controlling the temperature of the sample during SPME analysis of polynuclear aromatic hydrocarbons.

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Carc. 2 - Flam. Liq. 3 - STOT RE 1 - STOT RE 2

Target Organs

Central nervous system, hearing organs

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis of polynuclear aromatic hydrocarbons by SPME-GC-FID in environmental and tap waters.
Coelho, Emanuel, Celia Ferreira, and Cristina MM Almeida.
Journal of the Brazilian Chemical Society, 19, 1084-1097 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service