Skip to Content
Merck
All Photos(1)

Key Documents

56736

Supelco

Stainless Steel 1/16 in. Capillary Tubing

L × O.D. × I.D. 1.0 m × 1/16 in. × 0.030 in.

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$50.20

$50.20


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$50.20

About This Item

UNSPSC Code:
41115700
eCl@ss:
32119290

$50.20


Estimated to ship on01 June 2025


Request a Bulk Order

material

white stainless steel color band

L × O.D. × I.D.

1.0 m × 1/16 in. × 0.030 in.

Looking for similar products? Visit Product Comparison Guide

General description

316 stainless steel, 1/16 in. O.D. 1 m length is coiled, all other dimensions are straight.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service