Skip to Content
Merck
All Photos(1)

Key Documents

55050-U

RheBuild® Kit

for use with 8125/8126 valves

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$407.00

$407.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 EA
$407.00

About This Item

UNSPSC Code:
41115854
NACRES:
SB.51

$407.00


Check Cart for Availability

Request a Bulk Order

compatibility

for use with 8125/8126 valves

Looking for similar products? Visit Product Comparison Guide

General description

These kits include all of the tools and parts you need to repair your Rheodyne injector. The parts included are those most likely to be damaged, worn, or lost. Kits for front-loading injectors include: rotor seal, stator face assembly, isolation seal, needle guide, needle port cleaner, 2 hex keys, operating instructions, and a mini-manual. Parts may vary, depending on valve model.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

RheBuild is a registered trademark of IDEX Health & Science, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service