Skip to Content
Merck
All Photos(1)

Key Documents

54417

Rheodyne® 3-way Valve, 7030

Accepts 1/16″ tubing

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,410.00

$1,410.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
1 EA
$1,410.00

About This Item

UNSPSC Code:
41115700

$1,410.00


0 left in stock

Request a Bulk Order

material

stainless steel valve

description

manual 3-way switching valve

feature

number of ports 6
positions 2

manufacturer/tradename

Rheodyne

parameter

5000 psi max. pressure (350 bar) (factory set)

fitting

10-32 female Rheodyne valve ports

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Rheodyne is a registered trademark of IDEX Health & Science, LLC

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service