Skip to Content
Merck
All Photos(1)

Key Documents

54127-U

Supelco

2-(2-Pyridyl)ethyl Silica Gel

bed wt 100 mg, volume 1 mL, pk of 108

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
$594.00

$594.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
1 PKG
$594.00

About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

$594.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

composition

bed wt, 100 mg

packaging

pk of 108

technique(s)

solid phase extraction (SPE): suitable

volume

1 mL

matrix active group

WAX phase

application(s)

food and beverages

separation technique

ion exchange

General description

Retention Mechanism: Anion-exchange
Sample Matrix Compatibility: Organic or aqueous Solutions

  • Weak anion exchanger ideal for extracting strong basic compounds that remain charged at all pH levels
  • Unlike conventional weak anion exchange SPE phases such as -NH2 (aminopropyl) that have a pKa of 9-10, a pH ≤ 7 is required to protonate or ionize the stationary phase to facilitate analyte retention. Elution is typically done by increasing the pH to 11 resulting in neutralization of the SPE phase.
  • 2-(2-pyridyl)-ethyl silica gel has a pKa of ~6. Therefore, analyte elution is feasible at a pH ≥ 7. This characteristic is important for extracting analytes that are not stable (e.g. hydrolyzes) at high pHs typically required for elution when using traditional weak anion exchangers.
  • Ideal for extracting acyl-coenzyme A esters from tissue.
  • For more information, please see: Minkler, P.E., Kerner, J., Ingalls, S.T., Hoppel, C.L., Novel isolation procedure for short-, medium-, and long-chain acyl-coenzyme A esters from tissue, Analytical Biochemistry 376 (2008) 275–276

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrea Arias-Alvarado et al.
Analytical biochemistry, 615, 114067-114067 (2020-12-20)
Cellular availability of acetyl-CoA, a central intermediate of metabolism, regulates histone acetylation. The impact of a high-fat diet (HFD) on the turnover rates of acetyl-CoA and acetylated histones is unknown. We developed a method for simultaneous measurement of acetyl-CoA and

Protocols

Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service