Skip to Content
Merck
All Photos(2)

Key Documents

52797-U

Supelco

HybridSPE®-PL solid phase extraction (SPE) Cartridge

Bed wt. 100 mg, volume 3 mL, pk of 54 ×

Synonym(s):

HybridSPE®-Phospholipid

Sign Into View Organizational & Contract Pricing

Select a Size

54 EA
$474.00

$474.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Select a Size

Change View
54 EA
$474.00

About This Item

UNSPSC Code:
41115712
NACRES:
NB.21

$474.00


Check Cart for Availability

Request a Bulk OrderRequest a Sample

Product Name

HybridSPE-PL Cartridge, Bed wt. 100 mg, volume 3 mL, pk of 54 ×

material

PE frit (20/40 μm)
polypropylene hardware

Quality Level

description

Cartridge

composition

Bed wt., 100 mg

packaging

pk of 54 ×

technique(s)

solid phase extraction (SPE): suitable

volume

3 mL

matrix active group

zirconia-based phase

Looking for similar products? Visit Product Comparison Guide

General description

HybridSPE-Phospholipid technology is a simple and generic sample prep platform designed for the gross level removal of endogenous protein and phospholipid interferences from biological plasma and serum prior to LC-MS or LC-MS/MS analysis. Biological plasma or serum is first subjected to protein precipitation via the addition and mixing of acidified acetonitrile. Precipitated proteins are then removed by centrifugation and the resulting supernatant is loaded on the HybridSPE-Phospholipid 96-well plate or cartridge which acts as a chemical filter that specifically targets the removal of endogenous sample phospholipids. The phospholipid retention mechanism is based on a highly selective Lewis acid-base interaction between the proprietary zirconia ions functionally bonded to the HybridSPE-Phospholipid stationary phase and the phosphate moiety consistent with all phospholipids. The resulting eluent is ready for immediate LC-MS or LC-MS-MS analysis.

The "In-well" and "In-cartridge" precipitation methods are available for the HybridSPE-Phospholipid 96-well version and HybridSPE-Phospholipid Ultra cartridge in which biological plasma/serum is first added to either the well or cartridge, followed by acidified acetonitrile (precipitation agent). After a brief mixing/vortexing step, vacuum is applied. Because the 96-well and Ultra cartridge versions contain a series of low porosity hydrophobic filters/frits, the packed-bed filter/frit assembly acts as a depth filter facilitating the concurrent removal of both phospholipids and precipitated proteins during the extraction process. Standard HybridSPE-Phospholipid cartridges require an "off-line" precipitation method.

Features and Benefits

  • Merges the simplicity of protein precipitation and the selectivity of SPE via the targeted removal of phospholipids
  • Reduce ion-suppression through the complete removal of phospholipids and precipitated proteins
  • 2-3 step generic procedure
  • Minimal to no method development
  • Available in 96-well and 1 mL cartridge dimensions

Legal Information

HybridSPE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are Z-sep and HybridSPE different? If so, how?

    1 answer
    1. Both particles include a proprietary zirconia bonding and can be used in similar applications.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service