52653-U
Discovery® DSC-Si SPE Tube
bed wt. 100 mg, volume 1 mL, pk of 108
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
PE frit (20 μm porosity)
polypropylene hardware
Quality Level
composition
bed wt., 100 mg
packaging
pk of 108
technique(s)
solid phase extraction (SPE): suitable
surface area
480 m2/g
volume
1 mL
matrix
silica gel base material (irregularly shaped, acid washed)
matrix active group
silica bonding
particle size
50 μm
pore size
70 Å pore size
application(s)
food and beverages
separation technique
normal phase
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Retention Mechanism: Normal phase
Sample Matrix Compatibility: Organic solvents, oils, and lipids
Sample Matrix Compatibility: Organic solvents, oils, and lipids
- Unbonded acid washed silica sorbent ideal for normal phase SPE and other modified flash techniques
- Often used to separate or remove structurally similar molecules through successive elutions with increasingly polar solutions
- The most polar normal phase sorbent available
- Excellent capacity for purifying solution phase combinatorial chemistry reactions when removing target molecules from reaction by-products and excess reagents
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
11 - Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service