52574-U
Supelclean™ ENVI-Carb™-II/SAX/PSA SPE Tube
bed C 500 mg (Supelclean™ PSA SPE), bed B 500 mg (Supelclean™ LC-SAX SPE), bed A 500 mg (Supelclean™ ENVI-Carb-II SPE), volume 12 mL, pk of 20
About This Item
Recommended Products
material
PE frit (20 μm porosity)
polypropylene hardware
composition
bed A, 500 mg (Supelclean™ ENVI-Carb-II SPE)
bed B, 500 mg (Supelclean™ LC-SAX SPE)
bed C, 500 mg (Supelclean™ PSA SPE)
packaging
pk of 20
technique(s)
solid phase extraction (SPE): suitable
volume
12 mL
matrix active group
PSA phase
SAX phase
carbon phase
application(s)
food and beverages
separation technique
ion exchange
reversed phase
General description
Sample Matrix Compatibility: Organic or aqueous solutions
- Tri-layer SPE tube that contains Supelclean ENVI-Carb (upper layer), SAX (middle layer) & PSA (lower layer) SPE sorbents (separated by PE frit)
- Developed to offer superior clean up when conducting multi-residue pesticide analysis from food (e.g. agricultural products, meats, etc.).
- ENVI-carb has a strong affinity towards planar molecules, and can isolate/remove pigments (e.g., chlorophyll and carotinoids) and sterols commonly present in foods and natural products
- Supelclean PSA has a strong affinity and high capacity for fatty acids, organic acids, and some polar pigments and sugars
- Supelclean SAX offers additional ion-exchange exchange capacity for removing matrix components that may induce ion-suppression or enhancement during GC analysis.
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Reversed-phase interaction will retain most molecules with hydrophobic character; it is very useful for extracting analytes that are very diverse in structure within the same sample.
Protocols
Normal-phase SPE separates analytes based on polar interaction with sorbents in diverse sample matrices.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service