Key Documents
509744
SGE Syringes for PerkinElmer® Autosystem
volume 5 μL, 5F-PE-0.63, needle size 23 ga, needle L 70 mm, needle type, LTN (fixed)
Select a Size
1 EA
$218.00
$218.00
In StockDetails
Request a Bulk Order
Select a Size
Change View
1 EA
$218.00
About This Item
UNSPSC Code:
41122001
$218.00
In StockDetails
Request a Bulk Order
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
description
liquid syringe, fixed needle
feature
needle type LTN (fixed)
packaging
pkg of 1 ea
manufacturer/tradename
SGE 001954
needle L
70 mm
volume
5 μL
General description
SGE Syringes for PerkinElmer® Autosystem [volume 5 μL, 5F-PE-0.63, needle size 23 ga (cone tip), needle L 70 mm, needle type, LTN (fixed)] are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most trouble-free autosampler syringes available. The stainless steel cone tip, point style #1 is recommended for use with pre-drilled septa. The shape of this needle on the syringe has been developed for multiple and fast septum injections on the autosampler.
Legal Information
PerkinElmer is a registered trademark of PerkinElmer Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)