Skip to Content
Merck
All Photos(1)

Key Documents

50819-U

Supelco

Ascentis® Express 90 Å C18 (2 μm) HPLC Columns

L × I.D. 10 cm × 3 mm UHPLC Column

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,660.00

$1,660.00


Check Cart for Availability


Select a Size

Change View
1 EA
$1,660.00

About This Item

UNSPSC Code:
41115700
NACRES:
SB.52

$1,660.00


Check Cart for Availability

Product Name

Ascentis® Express C18, 2 μm UHPLC Column, 2 μm particle size, L × I.D. 10 cm × 3 mm

material

stainless steel column

Quality Level

Agency

suitable for USP L1

product line

Ascentis®

feature

endcapped

manufacturer/tradename

Ascentis®

packaging

1 ea of

parameter

1000 bar max. pressure (14500 psi)
60 °C temp. range

technique(s)

LC/MS: suitable
UHPLC-MS: suitable
UHPLC: suitable

L × I.D.

10 cm × 3 mm

surface area

120 m2/g

impurities

<5 ppm metals

matrix

Fused-Core particle platform
superficially porous particle

matrix active group

C18 (octadecyl) phase

particle size

2 μm

pore size

90 Å pore size

operating pH

2-9

application(s)

food and beverages

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pankaj Partani et al.
Journal of chromatographic science, 54(8), 1385-1396 (2016-05-27)
A liquid chromatography-tandem mass spectrometry method was developed and validated for the simultaneous determination of simvastatin (SV) and simvastatin acid (SVA) in human plasma. To improve assay sensitivity and achieve simultaneous analysis, SVA monitored in (-)ESI (electrospray ionization) mode within
Francesco Pio Prencipe et al.
Journal of chromatography. A, 1349, 50-59 (2014-05-27)
The study was aimed at developing a new analytical method for the metabolite fingerprinting of bioactive compounds in Humulus lupulus L. (hop), together with a simple extraction procedure. Different extraction techniques, including maceration, heat reflux extraction (HRE), ultrasound-assisted extraction (UAE)
Silvia Jakabová et al.
Journal of chromatography. A, 1232, 295-301 (2012-03-07)
Hyoscyamine (atropine) and scopolamine are the predominant tropane alkaloids in the Datura genus, occurring in all plant organs. The assessment of the alkaloid content of various plant parts is essential from the viewpoint of medical use, but also as a
Heena et al.
Journal of chromatographic science, 53(5), 800-806 (2014-10-18)
This study represents a new analytical high-performance liquid chromatography-fluorescence detector method for the determination of Al(III) as Al(III) complex with 8-hydroxyquinoline-5-sulfonic acid in a tap water sample and a coke sample. A micellar liquid chromatographic method is proposed for the
Ahmed Abrahim et al.
Journal of pharmaceutical and biomedical analysis, 51(1), 131-137 (2009-09-18)
Fused-core silica stationary phases represent a key technological advancement in the arena of fast HPLC separations. These phases are made by fusing a 0.5 microm porous silica layer onto 1.7 microm nonporous silica cores. The reduced intra-particle flow path of

Questions

  1. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service