Skip to Content
Merck
All Photos(1)

Key Documents

49055

Supelco

Parathion-methyl

PESTANAL®, analytical standard

Synonym(s):

O,O-Dimethyl O-(4-nitrophenyl) phosphorothioate, Methyl parathion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO5PS
CAS Number:
Molecular Weight:
263.21
Beilstein:
8905814
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

Quality Level

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 500 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

room temp

SMILES string

COP(=S)(OC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3

InChI key

RLBIQVVOMOPOHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Parathion-methyl is an organophosphate insecticide[1], which is extensively used for agricultural purposes.[2][3] Its mode of action involves the inactivation of the enzyme acetylcholinesterase (AChE) in insects and also mammals.[2] It undergoes degradation by soil microorganisms to yield p-nitrophenol as the product.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

114.8 °F - open cup

Flash Point(C)

46 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Absorption, distribution, metabolism and excretion of daily oral doses of [14 C] methyl parathion in hens
Abu-Qare WA, et al.
Toxicology Letters, 125(1), 1-10 (2001)
Isolation, characterization and identification of methyl parathion degrading bacteria from industrial waste
Vadukia, et al.
Journal of Cell and Tissue Research, 14(3), 4633-4633 (2014)
Effect of paraoxon-methyl and parathion-methyl on DNA in human lymphocytes and protective action of vitamin C
Blasiak J and Kowalik J
Pest Management Science, 55(12), 1182-1186 (1999)
Scott A Trammell et al.
Chemical communications (Cambridge, England), 48(34), 4121-4123 (2012-03-22)
Using a low power green laser, we have demonstrated a rate acceleration of ~2-fold for the hydrolysis of methyl parathion by irradiating the plasmon absorption band of Au nanoparticles capped with a Cu(bpy) catalyst.
Jingming Gong et al.
Biosensors & bioelectronics, 39(1), 320-323 (2012-08-08)
We developed a highly sensitive flow injection/amperometric biosensor for the detection of organophosphate pesticides (OPs) using layered double hydroxides (LDHs) as the immobilization matrix of acetylcholinesterase (AChE). LDHs provided a biocompatible microenvironment to keep the bioactivity of AChE, due to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service