Skip to Content
Merck
All Photos(1)

Key Documents

49042

Supelco

Heptachlor epoxide

PESTANAL®, analytical standard, isomer B

Synonym(s):

Heptachlor exo-epoxide, cis-Heptachlor epoxide, exo-1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-4,7-methano-3a,4,7,7a-tetrahydroindane, HCE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl7O
CAS Number:
Molecular Weight:
389.32
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

CofA

current certificate can be downloaded

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

Cl[C@H]1[C@H]2O[C@H]2[C@H]3[C@@H]1C4(Cl)C(Cl)=C(Cl)C3(Cl)C4(Cl)Cl

InChI

1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8?,9?/m0/s1

InChI key

ZXFXBSWRVIQKOD-TTWWGNBVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Heptachlor epoxide can be obtained as an oxidation product of heptachlor from soil microorganisms. It can find applications as an insecticide, basically used to control soil pests, which also includes termites.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Meador.PJ
Environmental Contaminants in Wildlife: Interpreting Tissue Concentrations (1996)
Montgomery.HJ
Groundwater Chemicals Field Guide (1991)
J P Hickey et al.
Archives of environmental contamination and toxicology, 50(1), 97-110 (2005-12-06)
Levels of chlorinated organic contaminants in predator fish have been monitored annually in each of the Great Lakes since the 1970s. This article updates earlier reports with data from 1991 to 1998 for lake trout (Salvelinus namaycush) and (Lake Erie
R J Smialowicz et al.
Toxicological sciences : an official journal of the Society of Toxicology, 61(1), 164-175 (2001-04-11)
This study was performed to determine if developmental exposure of rats to heptachlor (H) during the last half of gestation through puberty adversely affects adult functioning of the immune and reproductive systems. Time-bred pregnant female Sprague-Dawley rats were dosed by
William L Champion et al.
Journal of chromatography. A, 1024(1-2), 55-62 (2004-02-03)
Analytical high-performance liquid chromatographic separations of the individual enantiomers of five polychlorinated compounds were obtained on polysaccharide stereoselective HPLC columns. The enantiomers of the pesticides trans-chlordane, cis-chlordane and heptachlor were separated on CHIRALCEL OD using a hexane mobile phase. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service