Skip to Content
Merck
All Photos(1)

Key Documents

48996-U

Supelco

EPA 8151 Herbicide acid mix

in acetone (varied), PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$213.00

$213.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1 ML
$213.00

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

$213.00


Available to ship on22 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

Agency

EPA 8151

product line

PESTANAL®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

in acetone (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 8151 Herbicide acid mix is useful for identifying and quantifying the chlorinated herbicides on the EPA′s Appendix VIII and Appendix IX lists in solid wastes and groundwater at active hazardous treatment, storage, and disposal sites. The pre-requisite of EPA 8151 method is the use of gas chromatography (GC) with a selective detector or gas chromatography/mass spectrometry (GC/MS) or high performance liquid chromatography (HPLC).

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This product is now available as a certified reference material. Please reference CRM48996.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acifluorfen 100 μg/mL
    Bentazon 100 μg/mL
    Chloramben 100 μg/mL
    2,4-D 100 μg/mL
    2,4-DB acid 100 μg/mL
    Dalapon 100 μg/mL
    Tetrachloroterephthalic acid
    Dicamba 100 μg/mL
    3,5-Dichlorobenzoic acid 100 μg/mL
    Dichloroprop 100 μg/mL
    Dinoseb 100 μg/mL
    MCPA 10000 μg/mL
    MCPP 10000 μg/mL
    4-Nitrophenol 100 μg/mL
    Pentachlorophenol 100 μg/mL
    Picloram 100 μg/mL
    2,4,5-T 100 μg/mL
    2,4,5-TP 100 μg/mL
    See All (18)

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

1.4 °F

Flash Point(C)

-17.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service