Skip to Content
Merck
All Photos(1)

Key Documents

48705

Supelco

Aroclor 1221 solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 505,508,508A,8080,8081,8082,508.1
MISA (Canada) Group 27

product line

TraceCERT®

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT RE 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biodegradation of Aroclor 1221 type PCBs in sewage wastewater.
D Liu
Bulletin of environmental contamination and toxicology, 27(5), 695-703 (1981-11-01)
Adewale J Lambo et al.
Journal of basic microbiology, 46(2), 94-107 (2006-04-07)
A psychrotrophic bacterium isolated from polychlorinated biphenyls (PCBs)-contaminated soil grew on biphenyl as sole carbon and energy source, and actively cometabolized PCBs at low temperature. Analysis of cellular fatty acids indicate that the bacterium is most closely related to Hydrogenophaga
J Havel et al.
FEMS microbiology letters, 108(2), 211-217 (1993-04-01)
The hybrid Pseudomonas cepacia strain JHR22 was tested for its ability to degrade Aroclor 1221 in soil. The influence of supplements--mineral salts and trace elements--on the degradation was investigated. Disappearance of Aroclor 1221 congeners, occurrence of metabolites, and release of
F Fava
Chemosphere, 32(8), 1477-1483 (1996-04-01)
A bacterial co-culture, ECO3, constituted by a polychlorobiphenyl degrading bacterium, Pseudomonas sp. strain CPE1, and two chlorobenzoic acid degrading bacteria, could grow on Aroclor 1221 (75 mg/L) as the sole carbon source without accumulating chlorinated aromatic metabolites in the medium;
Selim Kutlu et al.
Cell biochemistry and function, 25(2), 167-172 (2005-09-24)
Polychlorinated biphenyls (PCBs) are persistent environmental pollutants. This study compared effects of two PCB mixtures, Aroclors 1221 (A1221) and 1254 (A1254) on serum levels of aspartate aminotransferase (AST), alanine aminotransferase (ALT), alkaline phosphatase (ALP), urea, creatinine and uric acid in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service