Skip to Content
Merck
All Photos(1)

Documents

48615

Supelco

Bromodichloromethane solution

certified reference material, 200 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CHBrCl2
CAS Number:
Molecular Weight:
163.83
EC Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 601
suitable for EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

200 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/CHBrCl2/c2-1(3)4/h1H

InChI key

FMWLUWPQPKEARP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S R Bielmeier et al.
Toxicology in vitro : an international journal published in association with BIBRA, 21(5), 919-928 (2007-03-09)
Bromodichloromethane (BDCM), a drinking water disinfection by-product, causes pregnancy loss, i.e. full-litter resorption, in F344 rats when treated during the luteinizing hormone (LH)-dependent period. This effect is associated with reduced maternal serum progesterone (P) and LH levels, suggesting that BDCM
National Toxicology Program genetically modified model report, (5)(5), 1-227 (2008-09-12)
Bromodichloromethane is a by-product of the chlorination of drinking water. It is formed by the halogen substitution and oxidation reactions of chlorine and naturally occurring organic matter (e.g., humic or fluvic acids) in water containing bromide. Bromodichloromethane was nominated to
Bromodichloromethane.
Report on carcinogens : carcinogen profiles, 12, 73-75 (2011-08-19)
National Toxicology Program technical report series, (532)(532), 1-248 (2006-06-03)
Bromodichloromethane is a by-product of the chlorination of drinking water. It is formed by the halogen substitution and oxidation reactions of chlorine with naturally occurring organic matter (e.g., humic or fulvic acids) in water containing bromide. Bromodichloromethane has been shown
Minh T Do et al.
Environmental health perspectives, 113(4), 418-424 (2005-04-07)
Chlorination disinfection by-products (CDBPs) are produced during the treatment of water with chlorine to remove bacterial contamination. CDBPs have been associated with an increased risk of bladder cancer. There is also some evidence that they may increase the risk of

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service