Skip to Content
Merck
All Photos(1)

Key Documents

48559

Supelco

Di-n-butyl phthalate

analytical standard, ampule of 5000 mg

Synonym(s):

Dibutyl phthalate, n-Butyl phthalate, DBP, Phthalic acid dibutyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-[CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
278.34
Beilstein:
1914064
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

9.6 (vs air)

vapor pressure

1 mmHg ( 147 °C)

autoignition temp.

756 °F

expl. lim.

0.47 %, 236 °F

packaging

ampule of 5000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.492 (lit.)

bp

340 °C (lit.)

mp

−35 °C (lit.)

density

1.043 g/mL at 25 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

SMILES string

CCCCOC(=O)c1ccccc1C(=O)OCCCC

InChI

1S/C16H22O4/c1-3-5-11-19-15(17)13-9-7-8-10-14(13)16(18)20-12-6-4-2/h7-10H,3-6,11-12H2,1-2H3

InChI key

DOIRQSBPFJWKBE-UHFFFAOYSA-N

Gene Information

mouse ... Esr1(13982)

Looking for similar products? Visit Product Comparison Guide

Application

Plasticizer

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

367.7 °F - open cup

Flash Point(C)

186.5 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carl Simonsson et al.
Toxicology and applied pharmacology, 264(1), 114-120 (2012-07-31)
Allergic contact dermatitis (ACD) is caused by low-molecular weight compounds called haptens. It has been shown that the potency of haptens can depend on the formulation in which they are applied on the skin. Specifically the sensitization potency of isothiocyanates
Alin C Dirtu et al.
Environment international, 59, 344-353 (2013-07-31)
Human exposure to chemicals commonly encountered in our environment, like phthalates, is routinely assessed through urinary measurement of their metabolites. A particular attention is given to the specific population groups, such as obese, for which the dietary intake of environmental
Harpreet Bhatia et al.
Environmental toxicology and chemistry, 32(10), 2335-2344 (2013-06-14)
Di-n-butyl phthalate (DnBP) is an industrial pollutant with antiandrogenic effects reported in male mammals and fish. Little research has been done on the endocrine effects of DnBP in female fish. The present study investigated the changes in ovarian histology and
Jun Wu et al.
Applied microbiology and biotechnology, 97(6), 2483-2491 (2012-06-26)
A dibutyl phthalate (DBP) transforming bacterium, strain M673, was isolated and identified as Acinetobacter sp. This strain could not grow on dialkyl phthalates, including dimethyl, diethyl, dipropyl, dibutyl, dipentyl, dihexyl, di(2-ethylhexyl), di-n-octyl, and dinonyl phthalate, but suspensions of cells could
Huamin Qiu et al.
Journal of pharmaceutical and biomedical analysis, 75, 123-129 (2012-12-19)
A microflow chemiluminescence (CL) sensor for determination of dibutyl phthalate (DBP) based on magnetic molecularly imprinted polymer (MMIP) as recognition element was fabricated. Briefly, a hydrophilic molecularly imprinted polymer layer was produced at the surface of Fe₃O₄@SiO₂ magnetic nanoparticles (MNPs)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service