Skip to Content
Merck
All Photos(1)

Key Documents

48540-U

Supelco

Bromodichloromethane

analytical standard

Synonym(s):

Dichlorobromomethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrCHCl2
CAS Number:
Molecular Weight:
163.83
Beilstein:
1697005
EC Number:
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.497 (lit.)

bp

87 °C (lit.)

mp

−55 °C (lit.)

density

1.98 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-8°C

SMILES string

ClC(Cl)Br

InChI

1S/CHBrCl2/c2-1(3)4/h1H

InChI key

FMWLUWPQPKEARP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bromodichloromethane (BDCM) is the most common trihalomethane (THM). It is a disinfection by-product identified in drinking water after chlorination, which appears to be carcinogenic and toxic. THMs are not only found in drinking supplies but are also prevalent in swimming pools, dump sites and fish samples.

Application

Bromodichloromethane may be used as an analytical standard for the determination of the analyte in drinking water and chlorinated seawater samples by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of trihalomethanes in chlorinated sea water samples using a purge-and-trap system coupled to gas chromatography
Allonier AS, et al.
Talanta, 51(3), 467-477 (2000)
Determination of trihalomethanes and some chlorinated solvents in drinking water by headspace technique with capillary column gas-chromatography
Kuivinen J and Johnsson H
Water Research, 33(5), 1201-1208 (1999)
Dose-dependent vehicle differences in the acute toxicity of bromodichloromethane
Lilly PD, et al.
Toxicological Sciences, 23(1), 132-140 (1994)
Bromodichloromethane, a trihalomethane that produces neoplasms in rodents
Dunnick JK, et al.
Cancer research, 47(19), 5189-5193 (1987)
Teresa L Leavens et al.
Toxicological sciences : an official journal of the Society of Toxicology, 99(2), 432-445 (2007-07-28)
Exposure to bromodichloromethane (BDCM), one of the most prevalent disinfection byproducts in drinking water, can occur via ingestion of water and by dermal absorption and inhalation during activities such as bathing and showering. The objectives of this research were to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service