Skip to Content
Merck
All Photos(1)

Key Documents

48230-U

Supelco

EPA 525 Fortification Solution A

certified reference material, 2000 μg/mL each component in dichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Agency

EPA 525

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type fortification

packaging

ampule of 1 mL

concentration

2000 μg/mL each component in dichloromethane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

EPA 525 Fortification Solution A is a multi-component solution containing acenaphthene-d10, chrysene-d12, and phenanthrene-d10 as certified reference materials (CRMs).
For use in extraction and quantification of general purpose organics in raw source water, finished drinking water, and drinking water at various stages of treatment.

Application

EPA 525 Fortification Solution A is used as a CRM to quantify polycyclic aromatic hydrocarbons (PAHs) in cigarettes and sidestream cigarette smoke by gas chromatography-mass spectrometry (GC-MS).[1] It may be used as a CRM in the microwave-assisted extraction of PAHs, polychlorinated biphenyls, phthalate esters, and nonylphenols from biota samples such as oysters, mussels, and fish liver[2] and sediment samples[3] before their quantification using GC-MS.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Acenaphthene-d10
    Chrysene-d12
    Phenanthrene-d10

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Characteristics of polycyclic aromatic hydrocarbon formation during smoking
Shimazu H, et al.
International Journal of GEOMATE, 14(42), 120-127 (2018)
Comparison of solid phase extraction, saponification and gel permeation chromatography for the clean-up of microwave-assisted biological extracts in the analysis of polycyclic aromatic hydrocarbons
Navarro P, et al.
International Journal of GEOMATE, 1128(1-2), 10-16 (2006)
Simultaneous microwave-assisted extraction of polycyclic aromatic hydrocarbons, polychlorinated biphenyls, phthalate esters and nonylphenols in sediments
Bartolome L, et al.
Journal of Chromatography A, 1068(2), 229-236 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service