Skip to Content
Merck
All Photos(1)

Key Documents

48161

Supelco

Biphenyl solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10
CAS Number:
Molecular Weight:
154.21
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 508,508.1

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C12H10/c1-3-7-11(8-4-1)12-9-5-2-6-10-12/h1-10H

InChI key

ZUOUZKKEUPVFJK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Yang Ou et al.
Chemical communications (Cambridge, England), 49(33), 3473-3475 (2013-03-22)
A novel A21-CuI catalyzed direct nitrogenation of biphenyl halides for the direct synthesis of carbazoles via a direct C-H amination process has been developed. A recyclable and inexpensive Cu-catalyst was successfully employed in N-heterocyclic compound synthesis via tandem azidation and
Erin M Wilfong et al.
Bioorganic & medicinal chemistry letters, 22(20), 6521-6524 (2012-09-19)
Fragment based drug discovery remains a successful tool for pharmaceutical lead discovery. Although based upon the principle of thermodynamic additivity, the underlying thermodynamic basis is poorly understood. A thermodynamic additivity analysis was performed using stromelysin-1 and a series of biphenyl
Aaron Fong et al.
Molecules (Basel, Switzerland), 18(2), 2281-2296 (2013-02-23)
Previous theoretical studies of Mislow's doubly-bridged biphenyl ketone 1 and dihydrodimethylphenanthrene 2 have determined significant entropic contributions to their normal (1) and inverse (2) conformational kinetic isotope effects (CKIEs). To broaden our investigation, we have used density functional methods to
Ondrej Uhlik et al.
PloS one, 7(7), e40653-e40653 (2012-07-19)
Bacteria were identified associated with biodegradation of aromatic pollutants biphenyl, benzoate, and naphthalene in a long-term polychlorinated biphenyl- and polyaromatic hydrocarbon-contaminated soil. In order to avoid biases of culture-based approaches, stable isotope probing was applied in combination with sequence analysis
Alan Fulp et al.
Journal of medicinal chemistry, 55(22), 10022-10032 (2012-10-27)
Cannabinoid receptor 1 (CB1) antagonists are potentially useful for the treatment of several diseases. However, clinical development of several CB1 antagonists was halted due to central nervous system (CNS)-related side effects including depression and suicidal ideation in some users. Recently

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service