Skip to Content
Merck
All Photos(1)

Key Documents

48088

Supelco

2-Bromo-1-chloropropane solution

certified reference material, 2000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6BrCl
CAS Number:
Molecular Weight:
157.44
EC Number:
MDL number:
UNSPSC Code:
41116107
Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 601

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type internal/surrogate

packaging

ampule of 1 mL

concentration

2000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

BrC(CCl)C

InChI

1S/C3H6BrCl/c1-3(4)2-5/h3H,2H2,1H3

InChI key

PUJJZGFFAQHYEX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Bromo-1-chloropropane may be used as an internal standard for the quantification of haloacetonitriles [1] and volatile organochlorine compounds in landfill leachates[2] using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup - Solvent

Flash Point(C)

11 °C - closed cup - Solvent

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Static headspace versus head space solid-phase microextraction (HS-SPME) for the determination of volatile organochlorine compounds in landfill leachates by gas chromatography
Menendez F C.J, et al.
Talanta, 63(4), 809-814 (2004)
Characterization of chlorination by-products in cooling effluents of coastal nuclear power stations
Allonier S-A, et al.
Marine Pollution Bulletin, 38(12), 1232-1241 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service