Skip to Content
Merck
All Photos(1)

Key Documents

47925

Supelco

Aroclor 1016 solution

certified reference material, 50 mg/kg in transformer oil

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 5 mL

concentration

50 mg/kg in transformer oil

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-30°C

InChI

1S/C12H7Cl3/c13-10-3-1-2-8(4-10)9-5-11(14)7-12(15)6-9/h1-7H

InChI key

RIBGNAJQTOXRDK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Aroclor 1016, a hazardous environmental contaminant, is a complex mixture of polychlorinated biphenyl (PCB) congeners, prepared by the chlorination of biphenyl.

Application

Aroclor 1016 may be used as a certified reference standard for the quantification of the analyte in environmental matrices using gas chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 1B - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

294.8 °F - closed cup

Flash Point(C)

146 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A disposable immunomagnetic electrochemical sensor based on functionalised magnetic beads and carbon-based screen-printed electrodes (SPCEs) for the detection of polychlorinated biphenyls (PCBs)
Centi.S, et al.
Analytica Chimica Acta, 538(1-2), 205-212 (2005)
Standard reference materials (SRMs) for the calibration and validation of analytical methods for PCBs (as Aroclor mixtures)
Poster.LD, et al.
Journal of research of the national institute of standards and technology, 109(2), 245-245 (2004)
R F Seegal et al.
Toxicology and applied pharmacology, 106(1), 136-144 (1990-10-01)
Exposure of the nonhuman primate, Macaca nemestrina, to Aroclor 1016, a commercial mixture of 26 lightly chlorinated PCB congeners, decreased dopamine concentrations in the caudate, putamen, substantia nigra, and hypothalamus. Only three ortho-substituted nonplanar PCB congeners (2,4,4', 2,4,2',4', and 2,5,2',5')
R F Seegal et al.
Toxicology, 66(2), 145-163 (1991-02-01)
Adult male non-human primates, Macaca nemestrina, were orally-exposed to corn oil or corn oil containing either Aroclor 1016 or 1260 at doses of 0.8, 1.6 or 3.2 mg/(kg.day) for 20 weeks. Brain concentrations of biogenic amines and individual PCB congeners
T H Ueng et al.
Toxicology, 35(2), 83-94 (1985-05-01)
The effects of the widely used industrial PCBs mixture, Aroclor 1016, as modifiers of monooxygenases were studied in rats and rabbits. From data presented, it is not possible to generalize the biological effects of PCBs observed with rats, namely, that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service