Skip to Content
Merck
All Photos(1)

Documents

47844

Supelco

Xylitol

analytical standard

Synonym(s):

Xylite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2[CH(OH)]3CH2OH
CAS Number:
Molecular Weight:
152.15
Beilstein:
1720523
EC Number:
MDL number:
UNSPSC Code:
12164500
PubChem Substance ID:
E Number:
E967

grade

analytical standard

CofA

current certificate can be downloaded

packaging

pkg of 1 g

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

94-97 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-30°C

SMILES string

OC[C@@H](O)[C@H](O)[C@@H](O)CO

InChI

1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+

InChI key

HEBKCHPVOIAQTA-SCDXWVJYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Xylitol is a 5-carbon sugar alcohol with antibacterial activity, found in medicines and dental products.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Xylitol may be used as a reference standard for the determination of xylitol in food samples by high performance liquid chromatography (HPLC) coupled to charged aerosol detector.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simultaneous separation and determination of erythritol, xylitol, sorbitol, mannitol, maltitol, fructose, glucose, sucrose and maltose in food products by high performance liquid chromatography coupled to charged aerosol detector.
Malgorzata G, et al.
Microchemical Journal, Devoted to the Application of Microtechniques in All Branches of Science, 117, 77-82 (2013)
Kiet A Ly et al.
Pediatric dentistry, 28(2), 154-163 (2006-05-20)
The purpose of this report was to provide an overview of xylitol and other polyol sweeteners and dental caries for clinicians and to discuss current applications for dental practice and potential community-based public health interventions. Xylitol, like other polyol sweeteners
Eun Joong Oh et al.
Metabolic engineering, 15, 226-234 (2012-10-30)
As Saccharomyces cerevisiae cannot utilize xylose as a carbon source, expression of XYL1 coding for xylose reductase (XR) from Scheffersomyces (Pichia) stipitis enabled production of xylitol from xylose with a high yield. However, insufficient supply of NAD(P)H for XR and
Lilian Raquel Hickert et al.
Bioresource technology, 143, 112-116 (2013-06-25)
Co-fermentation and simultaneous saccharification of rice hull hydrolysate (RHH) were investigated for the production of ethanol and xylitol by Saccharomyces cerevisiae, Spathaspora arborariae, or the combination of both. In bioreactor cultures under oxygen limitation, S. cerevisiae was capable of metabolizing
Xiaoxiao Guo et al.
Bioresource technology, 128, 547-552 (2012-12-06)
In this study, an integrated xylitol production pathway, directly using xylan as the substrate, was constructed in Candida tropicalis BIT-Xol-1 which could efficiently convert xylose into xylitol. In order to consolidate this bioprocessing, a β-1,4-xylanase gene (atn) and a β-xylosidase

Protocols

Separation of Maltotriose; Maltose; Xylitol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service