Skip to Content
Merck
All Photos(1)

Key Documents

47243

Supelco

1,3,5-Trinitrobenzene solution

certified reference material, 1000 μg/mL in acetonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3N3O6
CAS Number:
Molecular Weight:
213.10
UNSPSC Code:
12000000

grade

certified reference material
TraceCERT®

Agency

EPA 8330

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in acetonitrile

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H3N3O6/c10-7(11)4-1-5(8(12)13)3-6(2-4)9(14)15/h1-3H

InChI key

UATJOMSPNYCXIX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F

Flash Point(C)

2 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlos Medina et al.
The Journal of pathology, 224(4), 461-472 (2011-04-06)
Transforming growth factor β (TGF-β) is known to play a key role in intestinal fibrosis; however, the underlying mechanisms are not well understood. TGF-β signal transduction is through TGF-β receptors, including the TGF-β type 1 receptor. Most cell types contain
Barbara Chiavarino et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(33), 8185-8195 (2009-07-16)
Anion-pi interactions between a pi-acidic aromatic system and an anion are gaining increasing recognition in chemistry and biology. Herein, the binding features of an electron-deficient aromatic system (1,3,5-trinitrobenzene (TNB)) and selected anions (OH-, Br-, and I-) are examined in the
Guilherme R Lotufo et al.
Ecotoxicology and environmental safety, 73(5), 718-726 (2010-03-12)
Lethal effects of trinitrotoluene (TNT) to juvenile sheepshead minnows (JSHM) (Cyprinodon variegatus) were assessed in ten-day water exposures. Ten-day median lethal concentrations (LC50s) were 2.3 and 2.5 mg L(-1), the 10-d median lethal residue value (LR50) was 26.1 micromol kg(-1)
R W Nelson et al.
Bulletin of environmental contamination and toxicology, 84(1), 71-75 (2009-11-26)
The role of echinoid parental nutrition in early-life stage toxicity is not well understood. Arbacia punctulata were fed either a fresh diet consisting of organic lettuce and carrots or a dry feed. Embryos from parents fed the dry feed exhibited
Kent A Nielsen et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 17(39), 11001-11007 (2011-08-20)
The study of controllable molecular recognition in supramolecular receptors is important for elucidating design strategies that can lead to external control of molecular recognition applications. In this work, we present the design and synthesis of an asymmetric (TTF) tetrathiafulvalene-calix[4]pyrrole receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service