Skip to Content
Merck
All Photos(1)

Key Documents

46902-U

Supelco

cis-9-Octadecenoic acid methyl ester

certified reference material, 10 mg/mL in heptane

Synonym(s):

Oleic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

product line

TraceCERT®

form

liquid

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

10 mg/mL in heptane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

single component solution

functional group

ester

storage temp.

-10 to -25°C

InChI

1S/C19H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19(20)21-2/h10-11H,3-9,12-18H2,1-2H3/b11-10+

InChI key

QYDYPVFESGNLHU-ZHACJKMWSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Renee M Thomas et al.
Journal of the American Chemical Society, 133(19), 7490-7496 (2011-04-23)
N-Aryl,N-alkyl N-heterocyclic carbene (NHC) ruthenium metathesis catalysts are highly selective toward the ethenolysis of methyl oleate, giving selectivity as high as 95% for the kinetic ethenolysis products over the thermodynamic self-metathesis products. The examples described herein represent some of the
Ursula Biermann et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(26), 8201-8207 (2012-05-18)
A kinetic study of the dodecanethiol-catalyzed cis/trans isomerization of methyl oleate (cis-2) without added initiator was performed by focusing on the initiation of the radical chain reaction. The reaction orders of the rate of isomerization were 2 and 0.5 for
Michael A Rogers et al.
Langmuir : the ACS journal of surfaces and colloids, 25(15), 8556-8566 (2009-03-31)
The solvent type strongly affects the nucleation behavior of 12HSA and therefore strongly influences the peak nucleation rate, fiber length, spatial distribution of mass, and degree of branching. Using nonisothermal kinetic models, a correlation was established among the activation energy
L I Mazaletskaia et al.
Prikladnaia biokhimiia i mikrobiologiia, 46(2), 148-152 (2010-04-16)
Influence of the widely used food supplement lecithin on the effectiveness of the inhibiting effect of the natural antioxidants (quercitin, dihydroquercitin, and alpha-tocopherol) has been studied in dependence of the rate of free radicals generation in the model oxidation reactions.
Catherine M Klapperich et al.
Journal of biomedical materials research. Part A, 91(2), 378-384 (2008-11-05)
We describe a new class of biomaterials with potential for a variety of applications in tissue engineering, wound healing, and transdermal drug delivery. These materials are based on oleic methyl ester (OME), which is derived from various plant oils including

Protocols

Separation of Methyl decanoate; Methyl dodecanoate; Methyl myristate; Methyl palmitate; Methyl caprylate; Methyl oleate; Methyl linoleate; Methyl linolenate; Methyl stearate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service