Skip to Content
Merck
All Photos(1)

Key Documents

44690-U

Supelco

EPA 547 Glyphosate Solution

certified reference material, 1000 μg/mL in H2O

Synonym(s):

Glyphosate solution, N-(Phosphonomethyl)glycine

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
$72.40

$72.40


Estimated to ship on01 April 2025


Request a Bulk Order

Select a Size

Change View
1 ML
$72.40

About This Item

UNSPSC Code:
77101502
NACRES:
NA.24

$72.40


Estimated to ship on01 April 2025


Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 547

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

1000 μg/mL in H2O

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

multi-component solution

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

US EPA 500 Series methods are analytical methods for identifying and quantifying organic contaminants such as volatile organic compounds (VOCs), pesticides, synthetic organic compounds (SOCs), and trihalomethane disinfection byproducts (THMs) in raw source water, finished drinking water, and drinking water at all stages of treatment. For ISO/IEC 17025 accredited labs, it is important to show traceability of all results, so for calibration, certified reference materials (CRMs) must be used. Our rapidly expanding “TraceCERT” product range includes an increasing number of CRMs for pesticides. These products are manufactured according to ISO/IEC 17025 and ISO Guide 34 double accreditation, which is the highest achievable quality level for reference material producers and which is also referred to as “the gold standard”. The CRM solutions are produced gravimetrically using NIST traceable weights for balance calibration.

Application

EPA 547 Glyphosate Solution is used as a CRM to quantify the broad-spectrum herbicide glyphosate in beer samples by an electrochemical assay based on anti-glyphosate-antibody-modified magnetic beads and disposable electrochemical sensors.[1]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Glyphosate determination by coupling an immuno-magnetic assay with electrochemical sensors
Bettazzi F, et al.
Sensors, 18(9), 2965-2965 (2018)

Protocols

EPA Method 547 outlines the analysis of glyphosate in drinking water by direct aqueous injection HPLC, post column derivatization, and fluorescence detection

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service