Skip to Content
Merck
All Photos(2)

Key Documents

44420

Supelco

CERTAN® capillary bottle

4.5 ml bottle, diam. × H 16 mm × 71 mm

Synonym(s):

Capillary bottle CERTAN®

Sign Into View Organizational & Contract Pricing

Select a Size

5 EA
$206.00

$206.00


Available to ship on15 April 2025Details


Request a Bulk Order

Select a Size

Change View
5 EA
$206.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

$206.00


Available to ship on15 April 2025Details


Request a Bulk Order

material

clear glass vial

feature

closure type screw top vial

diam. × H

16 mm × 71 mm

volume

4.5 mL

Looking for similar products? Visit Product Comparison Guide

General description

HINT: To extract a solution from CERTAN® capillary vials use a syringe with a needle diameter of 0.8 mm or smaller and a minuimum 70 mm length.
These vials provide the advantage of a sealed ampule with the flexibility of a screw cap bottle or a septum vial. These unique, screw-cap sample containers with a 1.2 × 28mm height capillary opening are designed for the storage of volatile samples and reference standards.
       
Advantages:
  • No appreciable change in concentration -even when open
  • Secure storage, minimal evaporation
  • Minimizes contamination risk
  • Sample filling and removal using a standard GC syringe
  • Essentially spillproof
  • PTFE-lined screw-cap
         
Applications:
  • Storage of liquids removed from conventional glass ampules
  • Storage of stock and standard solutions
  • Storage of difficult samples such as volatile halogen compounds or BTEX aromatics
  • Archiving of production or QC samples and extracts

Application

To extract a solution from CERTAN® capillary vials use a syringe with a needle diameter of 0.8 mm or smaller and a minimum 70 mm length.

Legal Information

CERTAN is a registered trademark of LGC Group

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service