Skip to Content
Merck
All Photos(1)

Key Documents

442679

Supelco

Hexadecane

analytical standard

Synonym(s):

n-Hexadecane, Cetane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)14CH3
CAS Number:
Molecular Weight:
226.44
Beilstein:
1736592
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

vapor density

7.8 (vs air)

vapor pressure

1 mmHg ( 105.3 °C)

CofA

current certificate can be downloaded

autoignition temp.

395 °F

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.434 (lit.)

bp

287 °C (lit.)

mp

18 °C (lit.)

transition temp

solidification point 17.5-18.5 °C

density

0.773 g/mL at 25 °C (lit.)

application(s)

environmental
petroleum

format

neat

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCC

InChI

1S/C16H34/c1-3-5-7-9-11-13-15-16-14-12-10-8-6-4-2/h3-16H2,1-2H3

InChI key

DCAYPVUWAIABOU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Product 442679 has been discontinued. Product 52209 may be an acceptable replacement.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

275.0 °F - closed cup

Flash Point(C)

135 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jaejoon Jung et al.
Microbial ecology, 70(4), 912-921 (2015-05-10)
Red clay was previously used to enhance bioremediation of diesel-contaminated soil. It was speculated that the enhanced degradation of diesel was due to increased bacterial growth. In this study, we selected Acinetobacter oleivorans DR1, a soil-borne degrader of diesel and
Shweta Mishra et al.
Bioresource technology, 111, 148-154 (2012-03-13)
In the present study, n-hexadecane degradation in MSM was investigated by three bacteria identified as Pseudomonas aeruginosa PSA5, Rhodococcus sp. NJ2 and Ochrobactrum intermedium P2, isolated from petroleum sludge. During 10 days of incubation, n-hexadecane was degraded to 99% by
Homeira Ebrahimzadeh et al.
Talanta, 94, 77-83 (2012-05-23)
A simple and high sensitive technique based on three phase hollow fiber liquid phase microextraction (HF-LPME), optimized by using a four-variable experimental design and response surface methodology was performed to evaluate dextromethorphan hydrobromide (DEX) and chloropheniramine maleate (CLP) simultaneously in
Matthew A Churchward et al.
Biophysical journal, 94(10), 3976-3986 (2008-01-30)
The Ca(2+)-triggered merger of two apposed membranes is the defining step of regulated exocytosis. CHOL is required at critical levels in secretory vesicle membranes to enable efficient, native membrane fusion: CHOL-sphingomyelin enriched microdomains organize the site and regulate fusion efficiency
Diana Gutiérrez et al.
Applied and environmental microbiology, 81(10), 3336-3348 (2015-03-10)
Phage therapy is a promising option for fighting against staphylococcal infections. Two lytic phages, vB_SauM_phiIPLA-RODI (phiIPLA-RODI) and vB_SepM_phiIPLA-C1C (phiIPLA-C1C), belonging to the Myoviridae family and exhibiting wide host ranges, were characterized in this study. The complete genome sequences comprised 142,348

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service