Skip to Content
Merck
All Photos(1)

Key Documents

442614

Supelco

Hexaldehyde-2,4-DNPH

analytical standard

Synonym(s):

Hexanal 2,4-dinitrophenylhydrazone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=NNHC6H3(NO2)2
CAS Number:
Molecular Weight:
280.28
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

feature

standard type calibration

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

101-103 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

CCCCC\C=N\Nc1ccc(cc1[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C12H16N4O4/c1-2-3-4-5-8-13-14-11-7-6-10(15(17)18)9-12(11)16(19)20/h6-9,14H,2-5H2,1H3/b13-8+

InChI key

SJUYYAXVFFVQNX-MDWZMJQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denise Biagini et al.
Talanta, 216, 120979-120979 (2020-05-28)
Biomarkers of oxidative stress are generally measured in blood and its derivatives. However, the invasiveness of blood collection makes the monitoring of such chemicals during exercise not feasible. Saliva analysis is an interesting approach in sport medicine because the collection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service