Skip to Content
Merck
All Photos(1)

Key Documents

442521

Supelco

Chloropicrin

analytical standard

Synonym(s):

Trichloronitromethane, Chloropicrin, Nitrotrichloromethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3CNO2
CAS Number:
Molecular Weight:
164.38
Beilstein:
1756135
EC Number:
MDL number:
UNSPSC Code:
41121813
PubChem Substance ID:

grade

analytical standard

Quality Level

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.461 (lit.)

bp

112 °C (lit.)

density

1.657 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

[O-][N+](=O)C(Cl)(Cl)Cl

InChI

1S/CCl3NO2/c2-1(3,4)5(6)7

InChI key

LFHISGNCFUNFFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chloropicrin is a disinfection by-product that is commonly produced in chlorinated water. It is known to cause health problems in humans.[1]
Sigma-Aldrich offers chloropicrin for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analysis.

Application

Chloropicrin may be used as an analytical reference standard for the determination of the analyte in drinking water,[1] chemical warfare agents[2] and grain based products[3] by various chromatographic techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and use of mixed fumigant standards for multiresidue level determination by gas chromatography
Daft JL
Journal of Agricultural and Food Chemistry, 33(4), 563-566 (1985)
Retrospective identification of chemical warfare agents by high-temperature automatic thermal desorption-gas chromatography-mass spectrometry
Carrick WA, et al.
Journal of Chromatography A, 925(1-2), 241-249 (2001)
Application of different analytical methods for determination of volatile chlorination by-products in drinking water.
Nikolaou AD, et al.
Talanta, 56(4), 717-726 (2002)
Dongdong Yan et al.
Journal of agricultural and food chemistry, 60(20), 5023-5027 (2012-05-04)
Gelatin capsules containing chloropicrin (Pic gel cap) were developed as a new formulation to reduce the potential human exposure risks associated with injection application methods. The objective of this study was to test the efficacy of a Pic gel cap
Amisha D Shah et al.
Environmental science & technology, 45(8), 3657-3664 (2011-03-23)
The application of UV disinfection in water treatment is increasing due to both its effectiveness against protozoan pathogens, and the perception that its lack of chemical inputs would minimize disinfection byproduct formation. However, previous research has indicated that treatment of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service