Skip to Content
Merck
All Photos(1)

Key Documents

442425

Supelco

7,12-Dimethylbenz[a]anthracene

analytical standard

Synonym(s):

1,4-Dimethyl-2,3-benzophenanthrene, 9,10-Dimethyl-1,2-benzanthracene, DMBA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H16
CAS Number:
Molecular Weight:
256.34
Beilstein:
1912135
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

packaging

ampule of 100 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

122-123 °C (lit.)

application(s)

environmental

format

neat

storage temp.

room temp

SMILES string

Cc1c2ccccc2c(C)c3c1ccc4ccccc34

InChI

1S/C20H16/c1-13-16-8-5-6-9-17(16)14(2)20-18(13)12-11-15-7-3-4-10-19(15)20/h3-12H,1-2H3

InChI key

ARSRBNBHOADGJU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ramar Perumal Samy et al.
PloS one, 7(12), e48514-e48514 (2013-01-04)
Breast cancer is the most common cancer among women. To date, improvements in hormonal and cytotoxic therapies have not yet led to a sustained remission or cure. In the present study, we investigated the in vitro and in vivo antitumor
J DiGiovanni et al.
Drug metabolism reviews, 11(1), 61-101 (1980-01-01)
As the result of rapidly developing technological advances, our understanding of the biotransformation and bioactivation of 7, 12-DMBA has increased markedly in recent years. In terms of the metabolic conversion of this polynuclear aromatic hydrocarbon to reactive mutagen/carcinogens, the "bay
Anthony J Apostoli et al.
International journal of cancer, 134(5), 1055-1066 (2013-08-13)
Breast cancer is the leading cause of new cancer diagnoses among women. Using peroxisome proliferator-activated receptor (PPAR)γ((+/-)) mice, we showed normal expression of PPARγ was critical to stop 7,12-dimethylbenz[a]anthracene (DMBA)-induced breast tumorigenesis. PPARγ is expressed in many breast cell types
E Hallberg
Journal of biochemical toxicology, 5(2), 71-90 (1990-01-01)
The adrenal cortex contains high amounts of detoxifying enzymes, as well as generators and protectors of reactive oxygen species. The high content of cytochrome P-450 enzymes in the adrenal cortex together with its remarkable tendency to accumulate hydrophobic substances probably
Jennifer M Watson et al.
Lasers in surgery and medicine, 45(3), 155-166 (2013-01-31)
Ovarian cancer has an extremely high mortality rate resulting from poor understanding of the disease. In order to aid understanding of disease etiology and progression, we identify the endogenous fluorophores present in a mouse model of ovarian cancer and describe

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service