Skip to Content
Merck
All Photos(1)

Key Documents

442327

Supelco

2,6-Dimethylaniline

analytical standard

Synonym(s):

2,6-Xylidine, 2-Amino-1,3-dimethylbenzene, 2-Amino-m-xylene

Sign Into View Organizational & Contract Pricing

Select a Size

1000 MG
$33.90

$33.90


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
1000 MG
$33.90

About This Item

Linear Formula:
(CH3)2C6H3NH2
CAS Number:
Molecular Weight:
121.18
Beilstein:
636332
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

$33.90


Available to ship on22 April 2025Details


Request a Bulk Order

grade

analytical standard

Quality Level

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

>99% (GC)

CofA

current certificate can be downloaded

packaging

ampule of 1000 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.560 (lit.)

bp

214 °C/739 mmHg (lit.)

mp

10-12 °C (lit.)

density

0.984 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

storage temp.

2-30°C

SMILES string

Cc1cccc(C)c1N

InChI

1S/C8H11N/c1-6-4-3-5-7(2)8(6)9/h3-5H,9H2,1-2H3

InChI key

UFFBMTHBGFGIHF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dimethylaniline is a alpha2-adrenergic agonist metabolite of xylazine, which can find applications as a sedative agent in food producing animals.[1] It is also used in industries as a synthetic intermediate in the production of several products like dyes, pesticides and synthetic resins.[2]

Application

2,6-Dimethylaniline may be used as an analytical standard for the determination of the analyte in animal samples, human biological samples, milk samples and pharmaceutical preparations by chromatography and electroanalytical techniques.[3][4][5][6][7][8]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, Characterization, and Comparative 32P-Postlabeling Efficiencies of 2, 6-Dimethylaniline? DNA Adducts
Goncalves.LL, et al.
Chemical Research in Toxicology, 14, 165-174 (2001)
Tumor-promoting activity of 2, 6-dimethylaniline in a two-stage nasal carcinogenesis model in N-bis (2-hydroxypropyl) nitrosamine-treated rats
Koujitani T, et al.
Cancer Letters, 142, 161-171 (1999)
Determination and temperature effects of lidocaine (Lignocaine) hydrochloride, epinephrine, methylparaben, 2, 6-dimethylaniline, and p-hydroxybenzoic acid in USP Lidocaine Injection by ion-pair reversed-phase high pressure liquid chromatography
Smith DJ
Journal of Chromatographic Science, 19(5), 253-258 (1981)
Simultaneous determination of xylazine and its major metabolite, 2, 6-dimethylaniline, in bovine and swine kidney by liquid chromatography
Holland DC, et al.
Journal of AOAC (Association of Official Analytical Chemists) International, 76(4), 720-724 (1993)
Determination of local anaesthetics and their impurities in pharmaceutical preparations using HPLC method with amperometric detection
Fijalek Z, et al.
Journal of Pharmaceutical and Biomedical Analysis, 37(5), 913-918 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service