Skip to Content
Merck
All Photos(1)

Documents

40060

Supelco

N-Nitrosodiphenylamine solution

certified reference material, 5000 μg/mL in methanol

Synonym(s):

N-Nitroso-N-phenylaniline, Diphenylnitrosamine, Diphenylnitrosoamine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Quality Level

Agency

EPA 625,8270,Mar ′90, Dec ′90, Aug ′91 Statement of Work
EPA OLM04 (Statement of Work)

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C12H10N2O/c15-13-14(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

UBUCNCOMADRQHX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 3 - Flam. Liq. 2 - Skin Sens. 1 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junghoon Choi et al.
Water research, 36(4), 817-824 (2002-02-19)
Studies have been conducted specifically to investigate the hypothesis that N-nitrosodimethylamine (NDMA) can be produced by reactions involving monochloramine. Experiments were conducted using dimethylamine (DMA) as a model precursor. NDMA was formed from the reaction between DMA and monochloramine indicating
M Zielenska et al.
Mutation research, 202(1), 269-276 (1988-11-01)
The carcinogenic nitrosamines, N-nitrosomethylaniline (NMA) and N-nitrosodiphenylamine (NDphA), which have been previously reported negative or very weakly mutagenic in the Salmonella/microsome assay, were found to be mutagenic in the hisG428 Salmonella strain, TA104. NMA was moderately potent and NDphA was
The genetic toxicology of N-nitrosodiphenylamine.
D McGregor
Mutation research, 317(3), 195-211 (1994-06-01)
Masayoshi Sawamura et al.
Journal of agricultural and food chemistry, 53(10), 4281-4287 (2005-05-12)
The inhibitory effect of yuzu (Citrus junos Tanaka) essential oil on the formation of N-nitrosodimethylamine (NDMA) in the presence of vegetables (31 species) or saliva was investigated by HPLC. Most vegetable extracts enhanced the formation of NDMA. However, the formation
Oxidative remediation of diphenylamine in wastewater.
I S Jamall et al.
Bulletin of environmental contamination and toxicology, 76(4), 740-744 (2006-05-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service