Skip to Content
Merck
All Photos(1)

Documents

40054

Supelco

Nitrobenzene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

Agency

NIOSH 2005

product line

TraceCERT®

CofA

current certificate can be downloaded

feature

standard type calibration

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H5NO2/c8-7(9)6-4-2-1-3-5-6/h1-5H

InChI key

LQNUZADURLCDLV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - Repr. 1B - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F - closed cup

Flash Point(C)

11 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingang Huang et al.
Water research, 46(14), 4361-4370 (2012-06-19)
Sulfate is frequently found in wastewaters that contain nitrobenzene. To reveal the effect of sulfate on the reductive transformation of nitrobenzene to aniline--with acetate or propionate as potential electron donors in anaerobic systems--an acetate series (R1-R5) and a propionate series
Lu Zhaoyang et al.
Water science and technology : a journal of the International Association on Water Pollution Research, 66(8), 1799-1805 (2012-08-22)
The adsorption of phenol, p-nitrophenol, aniline, and nitrobenzene onto a commercial granular activated carbon (GAC: F400) preloaded with tannic acid (TA), a model background contaminant, was investigated. Compared with virgin GAC, the adsorption capacities of the four selected aromatic organic
Xiaohong Zhou et al.
Journal of environmental sciences (China), 24(5), 934-939 (2012-08-17)
Nitrobenzene is an important raw material and product, which presents a heavy threat to the ecosystem. The potential impacts of nitrobenzene on sediment oxygen demand (SOD) were studied in lake sediment simulating reactors receiving relatively low inputs of nitrobenzene. Oxygen
Yufan Zhang et al.
Analytica chimica acta, 752, 45-52 (2012-10-30)
Novel Pt nanoparticles (PN) ensemble on macroporous carbon (MPC) hybrid nanocomposites (PNMPC) were prepared through a rapidly and simple one-step microwave-assisted heating procedure. The obtained PNMPC was characterized by transmission electron microscopy (TEM), X-ray photoelectron spectroscopy (XPS), thermogravimetric analysis (TGA)
Jiping Jiang et al.
Journal of hazardous materials, 227-228, 280-291 (2012-06-06)
This paper presents a generic framework and decision tools of real-time risk assessment on Emergency Environmental Decision Support System for response to chemical spills in river basin. The generic "4-step-3-model" framework is able to delineate the warning area and the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service