33282
Vial Decapper, pliers-type
for use with 13mm crimp seals, pkg of 1 ea
Synonym(s):
Decapper
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
description
Decapper, pliers style
packaging
pkg of 1 ea
compatibility
for use with 13mm crimp seals
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Vial Decapper, pliers-type is used for quick and easy removal of a 13 mm aluminum seal.
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service