Skip to Content
Merck
All Photos(1)

Key Documents

33110-U

Supelco

Vials, crimp top, serum bottle

volume 100 mL, clear glass bottle, O.D. 51.7 mm × H (94.5 mm), pkg of 36 ea

Synonym(s):

Serum Bottle Crimp Vials

Sign Into View Organizational & Contract Pricing

Select a Size

36 EA
$145.00

$145.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
36 EA
$145.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

$145.00


Available to ship on22 April 2025Details


Request a Bulk Order

material

clear glass bottle

feature

autoclavable
closure type crimp top vial

packaging

pkg of 36 ea

O.D.

51.7 mm × H (94.5 mm)

volume

100 mL

fitting

thread for 20 mm

Looking for similar products? Visit Product Comparison Guide

General description

These vials are constructed of clear, borosilicate glass; closure not included.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Buenos días, Me gustaría saber que presión pueden aguantar estas botellas. Contando que la presión exterior es la presión atmosférica. Un saludo Sergio

    1 answer
    1. This product has not been tested for how much pressure it can withstand. Item Z742278 is a pressure-tested flask with a 1.5 bar max. pressure.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service