Skip to Content
Merck
All Photos(1)

Key Documents

29601-U

Hamilton® LC syringe for Waters WISP®

1702, volume 25 μL, for use with Waters, needle size (n/a)

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$324.00

$324.00


Estimated to ship on18 May 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$324.00

About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

$324.00


Estimated to ship on18 May 2025


Request a Bulk Order

material

glass body

needle size

(n/a)

feature

PTFE-tipped plungers

packaging

pkg of 1 ea

manufacturer/tradename

Hamilton 80020

needle L

n/a

volume

25 μL

compatibility

for use with Waters

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Hamilton is a registered trademark of Hamilton Co.
Waters is a registered trademark of Waters

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service