Skip to Content
Merck
All Photos(1)

Key Documents

29471-U

Supelco

Merlin MicroShot Injector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.03

packaging

pkg of 1 ea

manufacturer/tradename

Merlin Instrument 701-10

Looking for similar products? Visit Product Comparison Guide

General description

Improve your manual syringe technique instantly. The Merlin MicroShot Injector makes manual syringe injections convenient and reproducible using readily available chromatography syringes. The unique trigger mechanism makes each injection automatic when the needle is inserted into the injection port. The plunger displacement is fixed for precise volume delivery confirming reproducibility.

Features and Benefits

Features:
  • Fixed volume to eliminate sampling error
  • Five injection volumes available from 0.1μL to 2.0μL
  • Calibrated to NIST reference standards for traceable displacement accuracy
  • Quick and easy syringe replacement
  • One year warranty

Footnote

Syringe not included

Legal Information

MicroShot is a trademark of Merlin Instrument Co.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service