Skip to Content
Merck
All Photos(1)

Documents

28683-U

Supelco

Humonic Optiflow 520 Glassware Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41112500
NACRES:
SB.54

application(s)

environmental

General description

This kit includes: glassware (flow tube), squeeze bulb, and flexible tubing.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service