28340-U
SLB®-PAHms Capillary GC Column
L × I.D. 30 m × 0.25 mm, df 0.25 μm
About This Item
Recommended Products
material
fused silica
manufacturer/tradename
SLB®
parameter
25-350 °C temperature (isothermal)
25-360 °C temperature (programmed)
Beta value
250
df
0.25 μm
technique(s)
GC/MS: suitable
gas chromatography (GC): suitable
L × I.D.
30 m × 0.25 mm
column type
capillary polar
Looking for similar products? Visit Product Comparison Guide
General description
USP Code: None
Phase: Bonded; Proprietary
Temp. Limits: ambient to 350 °C (isothermal) or 360 °C (programmed)
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Study demonstrates SPME of BPA from an energy drink using an overcoated PDMS/DVB fiber, followed by GC/MS/MS analysis.
Protocols
A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.
Overcoated SPME fiber enables low-level BPA analysis in food via SPME-GC-MS/MS, meeting tighter migration limit regulations.
Paprika spice's PAH contamination risk due to environmental exposure highlights food safety concerns.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service