Skip to Content
Merck
All Photos(1)

Key Documents

27262

Supelco

Assembled screw cap with hole with PTFE/silicone septum

black polypropylene hole cap, thread for 8-425, PTFE/silicone, septum thickness 1.5 mm, for use with 2 mL vial (standard opening), pkg of 100 ea

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
$135.00

$135.00


Available to ship on22 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 EA
$135.00

About This Item

UNSPSC Code:
41122001

$135.00


Available to ship on22 April 2025Details


Request a Bulk Order

material

PTFE/silicone
black polypropylene hole cap

feature

closure type black polypropylene hole cap

packaging

pkg of 100 ea

septum thickness

1.5 mm

fitting

thread for 8-425

compatibility

for use with 2 mL vial (standard opening)

Looking for similar products? Visit Product Comparison Guide

Application

Assembled screw cap may be used to store fresh plant material for collecting volatiles using solid phase microextraction (SPME) technique.[1] It may also be used in liquid chromatography–tandem mass spectrometry (LC–MS/MS) method performed for determining bisphenol A, 4-t-octylphenol and 4-nonylphenol in human blood serum.[2]

Features and Benefits

The septa and closure was designed to provide a posiive physical lock to assure that preassembled components arrive intact.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bisphenol A, 4-t-octylphenol, and 4-nonylphenol determination in serum by Hybrid Solid Phase Extraction?Precipitation Technology technique tailored to liquid chromatography?tandem mass spectrometry.
Asimakopoulos, Alexandros G., and Nikolaos S. Thomaidis.
Journal of Chromatography. B, Analytical Technologies in the Biomedical and Life Sciences, 986, 85-93 (2015)
Electroantennographic responses of the small chestnut weevil Curculio sayi (Coleoptera: Curculionidae) to volatile organic compounds identified from chestnut reproductive plant tissue.
Keesey, Ian W., et al.
Environmental Entomology, 41.4, 933-940 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service