Key Documents
2637301
Supelco
Inlet Liner, Split/Splitless Type, Straight Design (packed with 10% OV-101 on 80/100 Chromosorb W HP)
pkg of 1 ea
About This Item
UNSPSC Code:
41115719
description
Split (wool packed)
packed
packaging
pkg of 1 ea
manufacturer/tradename
Varian 03-918956-00 (OEM)
L × O.D. × I.D.
54 mm × 5.0 mm × 3.4 mm
compatibility
configured for
for use with Varian 1078 Injector
General description
Deactivated (silanized) glass inlet liner for capillary injection ports.
  • Fits Varian (1078 and 1079 Injector)
Choose from one of the most recent versions:
Certificates of Analysis (COA)