Skip to Content
Merck
All Photos(2)

Key Documents

25084

Supelco

Empty Tube for Thermal Desorption

greener alternative

O.D. × L 1/4 in. × 3 1/2 in.

Synonym(s):

Empty Tube for PerkinElmer® ATD or TurboMatrix, Glass, pk 1

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$153.00

$153.00


Estimated to ship on13 April 2025


Request a Bulk Order

Select a Size

Change View
1 EA
$153.00

About This Item

UNSPSC Code:
41104008

$153.00


Estimated to ship on13 April 2025


Request a Bulk Order

material

glass tube

description

empty
without caps

packaging

pkg of 1 ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

O.D. × L

1/4 in. × 3 1/2 in.

application(s)

industrial hygiene

compatibility

for use with PerkinElmer

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. Thermal desorption tubes are reusable at least 50 to 100 times, and reduces the usage of accessories like transfer vials, caps, and pipette tips, thus aligns with “Waste Prevention”. Also, these tubes eliminate the usage of toxic solvents like CS2 to extract the VOCs of interest, and thus aligns with “Safer Solvents and Auxiliaries” principle. Click here for more information.

Legal Information

PerkinElmer is a registered trademark of PerkinElmer Inc.
TurboMatrix is a trademark of PerkinElmer Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service