Key Documents
24697-U
Supelco
Battery charger
universal charger, for use with 1060, 1062, 1063, & 1067 Air Samplers, AC/DC input 110 V / 240 V AC
About This Item
UNSPSC Code:
41104008
NACRES:
NB.24
Product 24697-U is not currently sold in your country. Contact Technical Service
technique(s)
whole air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
compatibility
for use with 1060, 1062, 1063, & 1067 Air Samplers
General description
Battery charger is a 110V/220V AC charger used as accessories for field sampling pumps used in air monitoring applications.
Choose from one of the most recent versions:
Certificates of Analysis (COA)